Merck
  • Tandem Knoevenagel-Michael addition of aryl sulfonimines with diethyl malonate for synthesis of arylidene dimalonates.
Tandem Knoevenagel-Michael addition of aryl sulfonimines with diethyl malonate for synthesis of arylidene dimalonates.
The Journal of organic chemistry (2007-06-26)
Renhua Fan, Weizi Wang, Dongming Pu, Jie Wu
ABSTRACT
A highly efficient, one-flask tandem Knoevenagel-Michael addition reaction of sulfonimines with diethyl malonate in the presence of a catalytic amount of base affords the corresponding arylidene dimalonates in good to excellent yields.
Sigma-Aldrich
Diethyl malonate, ReagentPlus®, 99%
Sigma-Aldrich
Diethyl malonate, ≥98%, FG