Merck
[Study of the pH sensitive polymer].
Guang pu xue yu guang pu fen xi = Guang pu (2006-10-25)
Liang-Qia Guo, Zeng-Hong Xie, Xu-Cong Lin, Xiang-Hua Zheng, Mao-Sheng Zhang, Guo-Nan Chen
ABSTRACT
A pH sensitive polymer was prepared by copolymerization of methacrylic acid as monomer, diethylene glycol dimethacrylate as cross-linking reagent, heptane as porogen, and fluorescent dye eosin as indicator. The factors of influence on the preparation, and the character of the pH sensitive polymer for pH were studied. The maximal emission wavelength of eosin was red shifted in the polymer than in solution, the apparent Ka largened, and the dissociation equilibrium of indicator was shifted to acidity direction, because the polarity of polymer diminished. Under the optimal condition, the calibration curve of the pH sensitive polymer covered the range of pH 0-3.0 with good reproduction and reversibility.
Sigma-Aldrich
Di(ethylene glycol) dimethacrylate, 95%