Merck
  • Fungal metabolic model for tyrosinemia type 3: molecular characterization of a gene encoding a 4-hydroxy-phenyl pyruvate dioxygenase from Aspergillus nidulans.
Fungal metabolic model for tyrosinemia type 3: molecular characterization of a gene encoding a 4-hydroxy-phenyl pyruvate dioxygenase from Aspergillus nidulans.
Eukaryotic cell (2006-08-10)
Márcia Eliana da Silva Ferreira, Marcela Savoldi, Pierina Sueli Bonato, Maria Helena S Goldman, Gustavo H Goldman
ABSTRACT
Mutations in the human HPD gene (encoding 4-hydroxyphenylpyruvic acid dioxygenase) cause hereditary tyrosinemia type 3 (HT3). We deleted the Aspergillus nidulans homologue (hpdA). We showed that the mutant strain is not able to grow in the presence of phenylalanine and that it accumulates increased concentrations of tyrosine and 4-hydroxyphenylpyruvic acid, mimicking the human HT3 phenotype.
Sigma-Aldrich
4-Hydroxyphenylpyruvic acid, 98%