Merck
  • Efficacy of ronidazole for treatment of feline Tritrichomonas foetus infection.
Efficacy of ronidazole for treatment of feline Tritrichomonas foetus infection.
Journal of veterinary internal medicine (2006-06-01)
Jody L Gookin, Christina N Copple, Mark G Papich, Matthew F Poore, Stephen H Stauffer, Adam J Birkenheuer, David C Twedt, Michael G Levy
ABSTRACT
To determine the efficacy of ronidazole (RDZ), tinidazole (TDZ), and metronidazole (MDZ) against Tritrichomonas foetus in vitro and of RDZ for treatment of feline naturally occurring or experimentally induced T. foetus infection. A cat naturally infected with T. foetus infection and diarrhea. Ten specific-pathogen-free (SPF) kittens. RDZ, TDZ, and MDZ were tested for activity against 3 different feline isolates of T. foetus in vitro. RDZ then was administered to a naturally infected cat at 10 mg/kg PO q24h for 10 days. SPF kittens were infected orogastrically with feline T. foetus and treated with either placebo or RDZ (10 mg/kg PO q12h for 14 days). Cats with relapsing infection or those receiving placebo were treated subsequently with RDZ (either 30 or 50 mg/kg PO q12h for 14 days). Feces were examined for T. foetus by direct microscopy, culture, and polymerase chain reaction (PCR) testing weekly. Both RDZ and TDZ killed T. foetus at concentrations >0.1 microg/mL in vitro. In the naturally infected cat, RDZ abolished diarrhea and T. foetus infection for 85 days after treatment, at which time infection and diarrhea relapsed. Retreatment with RDZ eradicated diarrhea and T. foetus infection for over 407 days. In experimentally induced infection, RDZ at 10 mg/kg caused initial improvement, but infection relapsed in all 5 cats 2 to 20 weeks after treatment. At 30 or 50 mg/kg, 10/10 cats were negative for T. foetus infection for follow-up durations of 21 to 30 weeks after treatment. Oral administration of RDZ at 30 to 50 mg/kg q12h for 14 days resolved diarrhea and eradicated infection (on the basis of polymerase chain reaction [PCR] testing) in 1 naturally infected cat and 10 experimentally inoculated cats receiving a different isolate of T. foetus.
Product Description
Supelco
Ronidazole, analytical standard