Merck
Synthesis and characterization of fluoro-substituted polyaniline.
Applied biochemistry and biotechnology (2002-01-11)
A L Sharma, M Gerard, R Singhal, B D Malhotra, S Annapoorni
ABSTRACT
Poly(2-fluoroaniline) was prepared by both chemical and electrochemical polymerization in acidic medium. Characterization of poly(2-fluoroaniline) was accomplished experimentally using ultraviolet-visible, Fourier transform infrared, differential scanning calorimetry, thermal gravimetric analysis, and X-ray diffraction techniques, respectively. Scanning electron microscopy studies revealed globular morphology of chemically synthesized poly(2-fluoroaniline). The cyclic voltammetric studies revealed diffusion-controlled phenomenon in electrochemically synthesized poly(2-fluoroaniline).
Product Description
Supelco
2-Fluoroaniline, ≥99%