Merck
  • Membrane androgen receptor activation induces apoptotic regression of human prostate cancer cells in vitro and in vivo.
Membrane androgen receptor activation induces apoptotic regression of human prostate cancer cells in vitro and in vivo.
The Journal of clinical endocrinology and metabolism (2004-12-09)
Anastassia Hatzoglou, Marilena Kampa, Christina Kogia, Ioannis Charalampopoulos, Panayiotis A Theodoropoulos, Ploutarchos Anezinis, Constantina Dambaki, Evangelia A Papakonstanti, Efstathios N Stathopoulos, Christos Stournaras, Achille Gravanis, Elias Castanas
ABSTRACT
Nongenomic androgen actions imply mechanisms different from the classical intracellular androgen receptor (iAR) activation. We have recently reported the identification of a membrane androgen receptor (mAR) on LNCaP human prostate cancer cells, mediating testosterone signal transduction within minutes. In the present study we provide evidence that activation of mAR by nonpermeable, BSA-coupled testosterone results in 1) inhibition of LNCaP cell growth (with a 50% inhibitory concentration of 5.08 nM, similar to the affinity of testosterone for membrane sites); 2) induction in LNCaP cells of both apoptosis and the proapoptotic Fas protein; and 3) a significant decrease in migration, adhesion, and invasion of iAR-negative DU145 human prostate cancer cells. These actions persisted in the presence of antiandrogen flutamide or after decreasing the content of iAR in LNCaP cells by iAR antisense oligonucleotides. Testosterone-BSA was also effective in inducing apoptosis of DU145 human prostate cancer cells, negative for iAR, but expressing mAR sites. In LNCaP cell-inoculated nude mice, treatment with testosterone-BSA (4.8 mg/kg body weight) for 1 month resulted in a 60% reduction of tumor size compared with that in control animals receiving only BSA, an effect that was not affected by the antiandrogen flutamide. Our findings suggest that activators of mAR may represent a new class of antitumoral agents of prostate cancer.
Sigma-Aldrich
Testosterone 3-(O-carboxymethyl)oxime: BSA - fluorescein isothiocyanate conjugate