Merck
  • [Impact of infusion therapy on the indices of endogenous intoxication in severe combined trauma of the abdominal organs].
[Impact of infusion therapy on the indices of endogenous intoxication in severe combined trauma of the abdominal organs].
Klinichna khirurhiia (2013-05-28)
V O Kryliuk
ABSTRACT
The peculiarities of course of endogenous intoxication (EI) in severe combined abdominal trauma were studied up in experiment. There was established, that in the laboratory animals, to whom the blood loss compensation was done, using colloid-hyperosmolar solution, the El severity was trustworthy lesser than in theothergroups.