Merck
  • Dynamics and function of ERM proteins during cytokinesis in human cells.
Dynamics and function of ERM proteins during cytokinesis in human cells.
FEBS letters (2017-09-11)
Shota Hiruma, Tomoko Kamasaki, Kohei Otomo, Tomomi Nemoto, Ryota Uehara
ABSTRACT
The molecular mechanism that governs cytoskeleton-membrane interaction during animal cytokinesis remains elusive. Here, we investigated the dynamics and functions of ERM (Ezrin/Radixin/Moesin) proteins during cytokinesis in human cultured cells. We found that ezrin is recruited to the cleavage furrow through its membrane-associated domain in a cholesterol-dependent but largely Rho-independent manner. While ERMs are dispensable for furrow ingression, they play a pivotal role in contractile activity of the polar cortex. Notably, when anillin and supervillin are codepleted, ERMs increasingly accumulate at the cleavage furrow and substantially contribute to the furrow ingression. These results reveal a supportive role of ERMs in cortical activities during cytokinesis, and also provide insight into the selective mechanism that preferentially associates cytokinesis-relevant proteins with the division site.
Sigma-Aldrich
Anti-alpha-Tubulin Antibody, clone YOL1/34, clone YOL1/34, from rat