Merck
  • Low-Dose Metformin Treatment Reduces In Vitro Growth of the LL/2 Non-small Cell Lung Cancer Cell Line.
Low-Dose Metformin Treatment Reduces In Vitro Growth of the LL/2 Non-small Cell Lung Cancer Cell Line.
Biomedicines (2023-01-22)
Nicole L Stott Bond, Didier Dréau, Ian Marriott, Jeanette M Bennett, Michael J Turner, Susan T Arthur, Joseph S Marino
ABSTRACT
Lung cancer maintains a relatively small survival rate (~19%) over a 5-year period and up to 80-85% of all lung cancer diagnoses are Non-Small Cell Lung Cancer (NSCLC). To determine whether metformin reduces non-small cell lung cancer (NSCLC) LL/2 cell growth, cells were grown in vitro and treated with metformin for 48 h. qPCR was used to assess genes related to cell cycle regulation and pro-apoptotic markers, namely Cyclin D, CDK4, p27, p21, and HES1. Treatment with 10 mM metformin significantly reduced HES1 expression (p = 0.011). Furthermore, 10 mM metformin treatment significantly decreased REDD1 (p = 0.0082) and increased p-mTOR Ser2448 (p = 0.003) protein expression. Control cells showed significant reductions in phosphorylated p53 protein expression (p = 0.0367), whereas metformin treated cells exhibited reduced total p53 protein expression (p = 0.0078). There were no significant reductions in AMPK, PKB/AKT, or STAT3. In addition, NSCLC cells were treated for 48 h. with 10 mM metformin, 4 µM gamma-secretase inhibitor (GSI), or the combination of metformin (10 mM) and GSI (4 µM) to determine the contribution of respective signaling pathways. Metformin treatment significantly reduced total nucleus expression of the proliferation maker Ki-67 with an above 65% reduction in Ki-67 expression between control and metformin-treated cells (p = 0.0021). GSI (4 µM) treatment significantly reduced Ki-67 expression by ~20% over 48 h (p = 0.0028). Combination treatment (10 mM metformin and 4 µM GSI) significantly reduced Ki-67 expression by more than 50% over 48 h (p = 0.0245). As such, direct administration of metformin (10 mM for 48 h) proved to be an effective pharmaceutical agent in reducing the proliferation of cultured non-small cell cancer cells. These intriguing in vitro results, therefore, support the further study of metformin in appropriate in vivo models as an anti-oncogenic agent and/or an adjunctive therapy.
Sigma-Aldrich
γ-Secretase Inhibitor X, InSolution, ≥90%, 1 mM