Merck
  • The Hippo pathway effector TAZ induces intrahepatic cholangiocarcinoma in mice and is ubiquitously activated in the human disease.
The Hippo pathway effector TAZ induces intrahepatic cholangiocarcinoma in mice and is ubiquitously activated in the human disease.
Journal of experimental & clinical cancer research : CR (2022-06-03)
Antonio Cigliano, Shanshan Zhang, Silvia Ribback, Sara Steinmann, Marcella Sini, Cindy E Ament, Kirsten Utpatel, Xinhua Song, Jingxiao Wang, Maria G Pilo, Fabian Berger, Haichuan Wang, Junyan Tao, Xiaolei Li, Giovanni M Pes, Serena Mancarella, Gianluigi Giannelli, Frank Dombrowski, Matthias Evert, Diego F Calvisi, Xin Chen, Katja Evert
ABSTRACT
Intrahepatic cholangiocarcinoma (iCCA) is a highly aggressive primary liver tumor with increasing incidence worldwide, dismal prognosis, and few therapeutic options. Mounting evidence underlines the role of the Hippo pathway in this disease; however, the molecular mechanisms whereby the Hippo cascade contributes to cholangiocarcinogenesis remain poorly defined. We established novel iCCA mouse models via hydrodynamic transfection of an activated form of transcriptional coactivator with PDZ-binding motif (TAZ), a Hippo pathway downstream effector, either alone or combined with the myristoylated AKT (myr-AKT) protooncogene, in the mouse liver. Hematoxylin and eosin staining, immunohistochemistry, electron microscopy, and quantitative real-time RT-PCR were applied to characterize the models. In addition, in vitro cell line studies were conducted to address the growth-promoting roles of TAZ and its paralog YAP. Overexpression of TAZ in the mouse liver triggered iCCA development with very low incidence and long latency. In contrast, co-expression of TAZ and myr-AKT dramatically increased tumor frequency and accelerated cancer formation in mice, with 100% iCCA incidence and high tumor burden by 10 weeks post hydrodynamic injection. AKT/TAZ tumors faithfully recapitulated many of the histomolecular features of human iCCA. At the molecular level, the development of the cholangiocellular lesions depended on the binding of TAZ to TEAD transcription factors. In addition, inhibition of the Notch pathway did not hamper carcinogenesis but suppressed the cholangiocellular phenotype of AKT/TAZ tumors. Also, knockdown of YAP, the TAZ paralog, delayed cholangiocarcinogenesis in AKT/TAZ mice without affecting the tumor phenotype. Furthermore, human preinvasive and invasive iCCAs and mixed hepatocellular carcinoma/iCCA displayed widespread TAZ activation and downregulation of the mechanisms protecting TAZ from proteolysis. Overall, the present data underscore the crucial role of TAZ in cholangiocarcinogenesis.
Sigma-Aldrich
Monoclonal Anti-WWTR1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0371, purified immunoglobulin, buffered aqueous glycerol solution