Merck
  • Sialylated and O-glycosidically linked glycans in prion protein deposits in a case of Gerstmann-Sträussler-Scheinker disease.
Sialylated and O-glycosidically linked glycans in prion protein deposits in a case of Gerstmann-Sträussler-Scheinker disease.
Neuropathology : official journal of the Japanese Society of Neuropathology (2010-07-30)
Viviana Zomosa-Signoret, Miguel Mayoral, Daniel Limón, Blanca Espinosa, Minerva Calvillo, Edgar Zenteno, Victor Martínez, Jorge Guevara
ABSTRACT
Prion diseases are caused by an abnormal form of the prion protein (PrP(Sc)). We identified, with lectins, post-translational modifications of brain proteins due to glycosylation in a Gerstmann-Sträussler-Scheinker (GSS) patient. The lectin Amaranthus leucocarpus (ALL), specific for mucin type O-glycosylated structures (Galß1,3 GalNAcα1,0 Ser/Thr or GalNAcα1,0 Ser/Thr), and Sambucus nigra agglutinin (SNA), specific for Neu5Acα2,6 Gal/GalNAc, showed positive labeling in all the prion deposits and in the core of the PrP(Sc) deposits, respectively, indicating specific distribution of O-glycosylated and sialylated structures. Lectins from Maackia amurensis (MAA, Neu5Acα2,3), Macrobrachium rosenbergii (MrL, Neu5,9Ac2-specific) and Arachis hypogaea (PNA, Gal-specific) showed low staining of prion deposits. Immunohistochemistry colocalization with prion antibody indicated that all lectins stained prion protein deposits. These results show that specific modifications in the glycosylation pattern are closely related to the hallmark lesions and might be an early event in neuronal degeneration in GSS disease.
Sigma-Aldrich
Anti-Prion Protein Antibody, NT, a.a. 78-97, serum, Chemicon®