- A concise and scalable route to L-azidohomoalanine.
A concise and scalable route to L-azidohomoalanine.
Nature protocols (2010-12-04)
Stefanie Roth, William C Drewe, Neil R Thomas
PMID21127494
ABSTRACT
A concise and highly efficient synthetic route to L-azidohomoalanine (L-Aha) and its homologues is presented here. These chemically modified amino acids are used for the introduction of bioorthogonal handles into proteins. The described route avoids major problems of previously reported methods including expensive starting materials, low efficiency, and lack of scalability. Starting from inexpensive N-Boc-O-Bn-L-aspartic acid, gram quantities of L-Aha hydrochloride can be prepared with high purity. The reactions can be completed within 1 week and the products can be incorporated into proteins using L-methionine auxotrophs.
Sigma-Aldrich
MAPK Pathway 1 Explorer Antibody MiniPack, clone AW39R, 1 mg/mL (05-538SP), Upstate®