Merck
  • Nicotinic acetylcholine receptors control acetylcholine and noradrenaline release in the rodent habenulo-interpeduncular complex.
Nicotinic acetylcholine receptors control acetylcholine and noradrenaline release in the rodent habenulo-interpeduncular complex.
British journal of pharmacology (2014-07-22)
F Beiranvand, C Zlabinger, A Orr-Urtreger, R Ristl, S Huck, P Scholze
ABSTRACT
Nicotinic acetylcholine receptors (nACh receptors) play a central role in the habenulo-interpeduncular system. We studied nicotine-induced release of NA and ACh in the habenula and interpeduncular nucleus (IPN). The habenula and IPN were loaded with [(3) H]-choline or [(3) H]-NA and placed in superfusion chambers. [(3) H]-ACh release was also stimulated using nicotinic agonists, electrical pulses and elevated [KCl]o in hippocampal and cortical slices from rats, wild-type mice and mice lacking α5, α7, β2, or β4 nACh receptor subunits. Finally, we analysed nACh receptor subtypes in the IPN using immunoprecipitation. Nicotine induced release of [(3) H]-ACh in the IPN of rats and mice. This release was calcium-dependent but not blocked by tetrodotoxin (TTX); moreover, [(3) H]-ACh release was abolished in β4-knockout mice but was unaffected in β2- and α5-knockout mice. In contrast, nicotine-induced release of [(3) H]-NA in the IPN and habenula was blocked by TTX and reduced in both β2-knockout and β4-knockout mice, and dose-response curves were right-shifted in α5-knockout mice. Although electrical stimuli triggered the release of both transmitters, [(3) H]-ACh release required more pulses delivered at a higher frequency. Our results confirm previous findings that β4-containing nACh receptors are critical for [(3) H]-ACh release in the mouse IPN. Experiments using α5-knockout mice also revealed that unlike in the hippocampus, nicotine-induced [(3) H]-NA release in the habenulo-interpeduncular system is altered in this knockout model. As α5-containing nACh receptors play a key role in nicotine intake, our results add NA to the list of transmitters involved in this mechanism.
Sigma-Aldrich
PANSORBIN® Cells, Standardized