Merck
  • One-Step Synthesis of Acidophilic Highly-Photoluminescent Carbon Dots Modified by Ionic Liquid from Polyethylene Glycol.
One-Step Synthesis of Acidophilic Highly-Photoluminescent Carbon Dots Modified by Ionic Liquid from Polyethylene Glycol.
ACS omega (2017-08-31)
Yujuan Chen, Qian Yang, Panpan Xu, Li Sun, Dong Sun, Kelei Zhuo
ABSTRACT
Acidophilic highly-photoluminescent ionic liquid (IL)-modified carbon dots (CDs) were fabricated directly from polyethylene glycol-2000 (PEG2000N) by a simple one-step hydrothermal method in a system containing an IL (1-butyl-3-methylimidazolium bromide [C4mim]Br) and hydrochloric acid (HCl). In this process, PEG2000N works as the carbon source, [C4mim]Br as the modifier, and HCl as the accelerator. CDs with low photoluminescence (PL) intensity and quantum yields (QYs) were generated in the system without H+, but CDs with high PL intensity and QYs could be prepared after H+ was introduced. Moreover, with the increase of H+ concentration, the QYs of the prepared CDs increase subsequently, and the highest QY reaches up to 43%. The formation mechanism was explored, and the results showed that H+ changes the surface groups of the CDs generated without H+ into those that exist on the CDs generated with H+, which further improves the PL performance of the CDs. Different from most CDs reported in the literature, the as-prepared CDs can still exhibit high PL intensity even under strong acidic condition.
Sigma-Aldrich
Polyethylene glycol, 2000, for synthesis