Merck
  • Scaffold-Hopping Approach To Discover Potent, Selective, and Efficacious Inhibitors of NF-κB Inducing Kinase.
Scaffold-Hopping Approach To Discover Potent, Selective, and Efficacious Inhibitors of NF-κB Inducing Kinase.
Journal of medicinal chemistry (2018-06-26)
Nicole Blaquiere, Georgette M Castanedo, Jason D Burch, Leonid M Berezhkovskiy, Hans Brightbill, Suzanne Brown, Connie Chan, Po-Chang Chiang, James J Crawford, Teresa Dong, Peter Fan, Jianwen Feng, Nico Ghilardi, Robert Godemann, Emily Gogol, Alice Grabbe, Alison J Hole, Baihua Hu, Sarah G Hymowitz, Moulay Hicham Alaoui Ismaili, Hoa Le, Patrick Lee, Wyne Lee, Xingyu Lin, Ning Liu, Paul A McEwan, Brent McKenzie, Hernani L Silvestre, Eric Suto, Swathi Sujatha-Bhaskar, Guosheng Wu, Lawren C Wu, Yamin Zhang, Zoe Zhong, Steven T Staben
ABSTRACT
NF-κB-inducing kinase (NIK) is a protein kinase central to the noncanonical NF-κB pathway downstream from multiple TNF receptor family members, including BAFF, which has been associated with B cell survival and maturation, dendritic cell activation, secondary lymphoid organ development, and bone metabolism. We report herein the discovery of lead chemical series of NIK inhibitors that were identified through a scaffold-hopping strategy using structure-based design. Electronic and steric properties of lead compounds were modified to address glutathione conjugation and amide hydrolysis. These highly potent compounds exhibited selective inhibition of LTβR-dependent p52 translocation and transcription of NF-κB2 related genes. Compound 4f is shown to have a favorable pharmacokinetic profile across species and to inhibit BAFF-induced B cell survival in vitro and reduce splenic marginal zone B cells in vivo.
Sigma-Aldrich
cUMP sodium salt, ≥98% (HPLC)