Merck
  • Inherited IFNAR1 Deficiency in a Child with Both Critical COVID-19 Pneumonia and Multisystem Inflammatory Syndrome.
Inherited IFNAR1 Deficiency in a Child with Both Critical COVID-19 Pneumonia and Multisystem Inflammatory Syndrome.
Journal of clinical immunology (2022-01-30)
Hassan Abolhassani, Nils Landegren, Paul Bastard, Marie Materna, Mohammadreza Modaresi, Likun Du, Maribel Aranda-Guillén, Fabian Sardh, Fanglei Zuo, Peng Zhang, Harold Marcotte, Nico Marr, Taushif Khan, Manar Ata, Fatima Al-Ali, Remi Pescarmona, Alexandre Belot, Vivien Béziat, Qian Zhang, Jean-Laurent Casanova, Olle Kämpe, Shen-Ying Zhang, Lennart Hammarström, Qiang Pan-Hammarström
ABSTRACT
Inborn errors of immunity (IEI) and autoantibodies to type I interferons (IFNs) underlie critical COVID-19 pneumonia in at least 15% of the patients, while the causes of multisystem inflammatory syndrome in children (MIS-C) remain elusive. To detect causal genetic variants in very rare cases with concomitant critical COVID-19 pneumonia and MIS-C. Whole exome sequencing was performed, and the impact of candidate gene variants was investigated. Plasma levels of cytokines, specific antibodies against the virus, and autoantibodies against type I IFNs were also measured. We report a 3-year-old child who died on day 56 of SARS-CoV-2 infection with an unusual clinical presentation, combining both critical COVID-19 pneumonia and MIS-C. We identified a large, homozygous loss-of-function deletion in IFNAR1, underlying autosomal recessive IFNAR1 deficiency. Our findings confirm that impaired type I IFN immunity can underlie critical COVID-19 pneumonia, while suggesting that it can also unexpectedly underlie concomitant MIS-C. Our report further raises the possibility that inherited or acquired dysregulation of type I IFN immunity might contribute to MIS-C in other patients.
Sigma-Aldrich
IFN-omega human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture