Merck
  • Determination of ametoctradin residue in fruits and vegetables by modified quick, easy, cheap, effective, rugged, and safe method using ultra-performance liquid chromatography/tandem mass spectrometry.
Determination of ametoctradin residue in fruits and vegetables by modified quick, easy, cheap, effective, rugged, and safe method using ultra-performance liquid chromatography/tandem mass spectrometry.
Food chemistry (2015-01-13)
Mingfeng Hu, Xingang Liu, Fengshou Dong, Jun Xu, Shasha Li, Hanqing Xu, Yongquan Zheng
ABSTRACT
A rapid, effective and sensitive method to quantitatively determine ametoctradin residue in apple, cucumber, cabbage, tomato and grape was developed and validated using ultra-performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS). The target compound was determined in less than 5.0 min using an electrospray ionisation source in positive mode (ESI+). The limit of detection was below 0.043 μg kg(-1), whereas the limits of quantification did not exceed 0.135 μg kg(-1) in all five matrices. The method showed excellent linearity (R(2)>0.9969) for the target compound. Recovery studies were performed in all matrices at three spiked levels (1, 10 and 100 μg L(-1)). The mean recoveries from five matrices ranged from 81.81% to 100.1%, with intra-day relative standard deviations (RSDr) in the range of 0.65-7.88% for the test compound. This method will be useful for the quick and routine detection of ametoctradin residues in potato, grape, cucumber, apple and tomato.
Product Description
Supelco
Ametoctradin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland