Merck
  • Enhancing the detection of Toxoplasma gondii via an anti-SAG1 scFv-alkaline phosphatase immunoconjugate.
Enhancing the detection of Toxoplasma gondii via an anti-SAG1 scFv-alkaline phosphatase immunoconjugate.
Biotechnology reports (Amsterdam, Netherlands) (2019-07-26)
Emna Hannachi, Aïda Bouratbine, Mohamed Mousli
ABSTRACT
The purpose of this study was to implement a fluorometric method for enhancing the detection sensitivity of Toxoplasma gondii in biological fluids. To address this challenge, we designed and produced a recombinant immunoconjugate tool based on a single-chain antibody fragment anti-T. gondii SAG1 antigen (scFvSG15) genetically fused to the bacterial alkaline phosphatase (AP) using 4-methyl-umbelliferyl-phosphate as fluorogenic substrate. The anti-SAG1 scFv-AP conjugate was fully bifunctional and was used successfully in different assays including immunoblot, fluorometric ELISA and direct immunofluorescence. The fluorometric immunoassay afforded an extremely low detection limit (1 tachyzoite/well), which was in agreement with the real-time PCR control test. The immunofluorescence imaging has provided captivating visual evidence of T. gondii detection. These results strongly suggest that the recombinant anti-SAG1-AP conjugate generated here might serve as useful and highly sensitive immunoassay probe to direct detect T. gondii in a one-step procedure, opening up new perspectives for diagnosis of toxoplasmosis.
Sigma-Aldrich
4-Methylumbelliferyl phosphate, phosphatase substrate