Merck
  • Effective Menin inhibitor-based combinations against AML with MLL rearrangement or NPM1 mutation (NPM1c).
Effective Menin inhibitor-based combinations against AML with MLL rearrangement or NPM1 mutation (NPM1c).
Blood cancer journal (2022-01-13)
Warren Fiskus, Steffen Boettcher, Naval Daver, Christopher P Mill, Koji Sasaki, Christine E Birdwell, John A Davis, Koichi Takahashi, Tapan M Kadia, Courtney D DiNardo, Qi Jin, Yuan Qi, Xiaoping Su, Gerard M McGeehan, Joseph D Khoury, Benjamin L Ebert, Kapil N Bhalla
ABSTRACT
Treatment with Menin inhibitor (MI) disrupts the interaction between Menin and MLL1 or MLL1-fusion protein (FP), inhibits HOXA9/MEIS1, induces differentiation and loss of survival of AML harboring MLL1 re-arrangement (r) and FP, or expressing mutant (mt)-NPM1. Following MI treatment, although clinical responses are common, the majority of patients with AML with MLL1-r or mt-NPM1 succumb to their disease. Pre-clinical studies presented here demonstrate that genetic knockout or degradation of Menin or treatment with the MI SNDX-50469 reduces MLL1/MLL1-FP targets, associated with MI-induced differentiation and loss of viability. MI treatment also attenuates BCL2 and CDK6 levels. Co-treatment with SNDX-50469 and BCL2 inhibitor (venetoclax), or CDK6 inhibitor (abemaciclib) induces synergistic lethality in cell lines and patient-derived AML cells harboring MLL1-r or mtNPM1. Combined therapy with SNDX-5613 and venetoclax exerts superior in vivo efficacy in a cell line or PD AML cell xenografts harboring MLL1-r or mt-NPM1. Synergy with the MI-based combinations is preserved against MLL1-r AML cells expressing FLT3 mutation, also CRISPR-edited to introduce mtTP53. These findings highlight the promise of clinically testing these MI-based combinations against AML harboring MLL1-r or mtNPM1.
Sigma-Aldrich
Anti-BCL2A1/BFL-1 Antibody, from rabbit, purified by affinity chromatography