Merck
  • Resampling improves the efficiency of a "fast-switch" equilibrium sampling protocol.
Resampling improves the efficiency of a "fast-switch" equilibrium sampling protocol.
The Journal of chemical physics (2009-03-05)
Edward Lyman, Daniel M Zuckerman
ABSTRACT
We recently applied a multistage reweighting scheme to demonstrate the sampling of equilibrium configurational distributions of peptides from nonequilibrium, simulated annealing trajectories [E. Lyman and D. M. Zuckerman, J. Chem. Phys. 127, 065101 (2007)]. Here we demonstrate that a statistical variance reduction technique, resampling, improves the efficiency of the protocol by about a factor of 3 in a penta-alanine system. While we are not optimistic for the ultimate efficiency of purely temperature-based sampling methods, resampling-type improvements ought to find application in other (formally equivalent) nonequilibrium sampling protocols, such as Jarzynski-relation calculations and annealing-based NMR structure calculations.
Sigma-Aldrich
Ala-Ala-Ala-Ala-Ala