Merck
Kinases of the Src family: structure and functions.
Biochemistry. Biokhimiia (2000-03-07)
A G Tatosyan, O A Mizenina
ABSTRACT
Tyrosine kinases of the Src family are involved in different signal transduction pathways in cells. The corresponding genes participate in such vital processes as growth, differentiation, adhesion, transcription, etc. Specific structural changes confer oncogenic properties to the Src protein. In this review, we summarize the available data on the structure, substrates, regulation mechanisms, and role of nonreceptor tyrosine kinases by the example of the src gene product (as the prototype member of this family) and a number of related proteins.
Sigma-Aldrich
FGR, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution