Merck
  • Development of eco-friendly CO2-responsive cellulose nanofibril aerogels as "green" adsorbents for anionic dyes removal.
Development of eco-friendly CO2-responsive cellulose nanofibril aerogels as "green" adsorbents for anionic dyes removal.
Journal of hazardous materials (2020-10-25)
Lin Yang, Yifei Zhan, Yujia Gong, Erhui Ren, Jianwu Lan, Ronghui Guo, Bin Yan, Sheng Chen, Shaojian Lin
ABSTRACT
A novel CO2-responsive cellulose nanofibril aerogel as a "green" adsorbent derived from poly(methacrylic acid-co-2-(dimethylamino) ethyl methacrylate) and carboxylated cellulose nanofibrils was successfully prepared via stepwise cation-induced gelation and freeze drying method. This aerogel exhibited CO2-triggered adsorption behavior towards anionic dyes with a rapid adsorption rate and a high adsorption capacity, as well as satisfactory mechanical properties. Upon CO2 stimulation, the charged aerogel can selectively adsorb anionic dyes from aqueous solutions based on an electrostatic interaction. The maximum adsorption capacities of this aerogel towards methyl blue (MB), naphthol green B (NGB), and methyl orange (MO) were 598.8, 621.1 and 892.9 mg g-1, respectively, accompanied by fast adsorption equilibriums towards MB and NGB within 7 min, and MO within 12 min. Meanwhile, the adsorption isotherms and the kinetics of the CO2-responsive adsorbents followed the Freundlich isotherm and the pseudo-second-order model, respectively. Furthermore, the resulting CO2-responsive adsorbent exhibited outstanding recyclability, as its adsorption performance can still be maintained even after twenty cycles. Accordingly, the resultant CO2-responsive cellulose nanofibril aerogel could be a promising adsorbent material for the removal of anionic dyes in wastewater remediation.