Merck
  • A modified two-color flow cytometry assay to quantify in-vitro reinvasion and determine invasion phenotypes at low Plasmodium falciparum parasitemia.
A modified two-color flow cytometry assay to quantify in-vitro reinvasion and determine invasion phenotypes at low Plasmodium falciparum parasitemia.
Experimental parasitology (2020-08-29)
Ines Atuh Ngoh, Damian Nota Anong, Jerome Cho Fru, Fatoumata Bojang, Haddijatou Mbye, Alfred Amambua-Ngwa
ABSTRACT
Invasion of human red blood cells (RBCs) by Plasmodium parasites is a crucial yet poorly characterised phenotype. Two-color flow cytometry (2cFCM) promises to be a very sensitive and high throughput method for phenotyping parasite invasion. However, current protocols require high (~1.0%) parasitemia for assay set-up and need to be adapted for low parasitemia samples, which are becoming increasingly common in low transmission settings. Background fluorescence from nuclei-containing uninfected RBCs and high autologous reinvasion rates (merozoite invasion of donor uninfected RBCs present at 50% assay volume) are some of the limitations to the method's sensitivity to enumerate low parasitemia (<0.5%) with nucleic acid-based stains. Here, we describe modifications for plating unlabeled donor to labeled target RBCs per assay well and for gating parasitemia, that produces accurate quantifications of low reinvasion parasitemia. Plasmodium falciparum 3D7, Dd2 and field isolates at various low and high parasitemia (0.05%-2.0%) were used to set-up SyBr Green 1-based 2cFCM invasion assays. Target RBCs were labeled with CTFR proliferation dye. We show that this dye combination allowed for efficient parasite invasion into target RBCs and that a 1:3 ratio of unlabeled to labeled RBCs per assay greatly skewed autologous reinvasion (p < 0.001). Accuracy of quantifying reinvasion was limited to an assay parasitemia of 0.02% with minimal background interference. Invasion inhibition by enzymatic treatments increased averagely by 10% (p<0.05) across the entire parasitemia range. The effect was greater for samples with <0.5% parasitemia. Overall, a more sensitive method for phenotyping invasion of low P. falciparum parasitemia is described.
Sigma-Aldrich
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein