Merck
Stress-free automatic sleep deprivation using air puffs.
Journal of neuroscience methods (2015-05-28)
Brooks A Gross, William M Vanderheyden, Lea M Urpa, Devon E Davis, Christopher J Fitzpatrick, Kaustubh Prabhu, Gina R Poe
ABSTRACT
Sleep deprivation via gentle handling is time-consuming and personnel-intensive. We present here an automated sleep deprivation system via air puffs. Implanted EMG and EEG electrodes were used to assess sleep/waking states in six male Sprague-Dawley rats. Blood samples were collected from an implanted intravenous catheter every 4h during the 12-h light cycle on baseline, 8h of sleep deprivation via air puffs, and 8h of sleep deprivation by gentle handling days. The automated system was capable of scoring sleep and waking states as accurately as our offline version (∼90% for sleep) and with sufficient speed to trigger a feedback response within an acceptable amount of time (1.76s). Manual state scoring confirmed normal sleep on the baseline day and sleep deprivation on the two manipulation days (68% decrease in non-REM, 63% decrease in REM, and 74% increase in waking). No significant differences in levels of ACTH and corticosterone (stress hormones indicative of HPA axis activity) were found at any time point between baseline sleep and sleep deprivation via air puffs. There were no significant differences in ACTH or corticosterone concentrations between sleep deprivation by air puffs and gentle handling over the 8-h period. Our system accurately detects sleep and delivers air puffs to acutely deprive rats of sleep with sufficient temporal resolution during the critical 4-5h post learning sleep-dependent memory consolidation period. The system is stress-free and a viable alternative to existing sleep deprivation techniques.
Product Description
Millipore
MILLIPLEX® Rat Stress Hormone Magnetic Bead Panel - Endocrine Multiplex Assay, This MILLIPLEX MAP Rat Stress Hormone Magnetic Bead Panel is an Endocrine multiplex assay. The analytes available for this multiplex kit are: ACTH, Corticosterone, Melatonin.