Merck
  • Adrenal gland-dependent augmentation of plasminogen activator inhibitor-1 expression in streptozotocin-induced diabetic mice.
Adrenal gland-dependent augmentation of plasminogen activator inhibitor-1 expression in streptozotocin-induced diabetic mice.
Journal of thrombosis and haemostasis : JTH (2006-07-15)
K Oishi, N Ohkura, N Ishida
ABSTRACT
Diabetes is associated with an excess risk of cardiac events, and one risk factor for infarction is an elevated level of plasminogen activator inhibitor-1 (PAI-1). To evaluate whether the glucocorticoid hormones are involved in the diabetes-induced PAI-1 production, we examined expression profiles of PAI-1 mRNA in adrenalectomized (ADX) mice with streptozotocin (STZ)-induced diabetes. The diabetes-induced augmentation of plasma PAI-1 levels and PAI-1 mRNA expression in the heart and lungs was completely normalized in diabetic ADX mice. The glucocorticoid receptor antagonist RU486 significantly, but only partly suppressed PAI-1 induction in STZ-induced diabetic mice, suggesting that factors other than glucocorticoids are also involved in PAI-1 induction provoked by diabetes. Our results suggested that the adrenal gland plays a critical role in the progression of thrombosis in diabetic patients by inducing expression of the PAI-1 gene.
Sigma-Aldrich
GenElute mRNA Miniprep Kit, sufficient for 70 purifications