- Understanding allosteric interactions in hMLKL protein that modulate necroptosis and its inhibition.
Understanding allosteric interactions in hMLKL protein that modulate necroptosis and its inhibition.
Mixed Lineage Kinase domain-Like (MLKL), a key player in necroptosis, is a multi-domain protein with an N-terminal 4 helical bundle (4HB) and a pseudokinase domain (PsK) connected by brace helices. Phosphorylation of PsK domain of MLKL is a key step towards oligomerization of 4HB domain that causes cell death. Necrosulfonamide (NSA) binds to the 4HB domain of MLKL to inhibit necroptosis. To understand the molecular details of MLKL function and it's inhibition, we have performed a molecular dynamic study on hMLKL protein in apo, phosphorylated and NSA-bound states for a total 3 μs simulation time. Our simulations show increased inter-domain flexibility, increased rigidification of the activation loop and increased alpha helical content in the brace helix region revealing a form of monomeric hMLKL necessary for oligomerization upon phosphorylation as compared to apo state. NSA binding disrupts this activated form and causes two main effects on hMLKL conformation: (1) locking of the relative orientation of 4HB and PsK domains by the formation of several new interactions and (2) prevention of key 4HB residues to participate in cross-linking for oligomer formation. This new understanding of the effect of hMLKL conformations on phosphorylation and NSA binding suggest new avenues for designing effective allosteric inhibitors of hMLKL.