Merck
  • Identification of (+)-gallocatechin as a bio-antimutagenic compound in Psidium guava leaves.
Identification of (+)-gallocatechin as a bio-antimutagenic compound in Psidium guava leaves.
Phytochemistry (1994-07-01)
T Matsuo, N Hanamure, K Shimoi, Y Nakamura, I Tomita
ABSTRACT
From the MeOH-extract of guava leaves, (+)-gallocatechin was isolated as a bio-antimutagenic compound against UV-induced mutation in Escherichia coli. This strengthens the evidence that phenolic compounds require three neighbouring-OH groups in order to possess this activity.
Sigma-Aldrich
(−)-Gallocatechin, ≥98% (HPLC)