Merck
  • Rethinking Schizophrenia and Depression Comorbidity as One Psychiatric Disorder Entity: Evidence From Mouse Model.
Rethinking Schizophrenia and Depression Comorbidity as One Psychiatric Disorder Entity: Evidence From Mouse Model.
Frontiers in neuroscience (2020-03-29)
Chunhua Zhou, Dezhi Kong, Xiaodong Zhu, Wei Wu, Rong Xue, Gongying Li, Yong Xu, Sha Liu, Hongjun Tian, Chuanjun Zhuo
ABSTRACT
Schizophrenia is frequently accompanied by depressive symptoms, but the pathological mechanisms remain to be elucidated. In this study, we used chronic unpredicted mild stress plus MK801 injection to generate a mouse model of schizophrenia with depression, in which in vivo 2-photon calcium imaging and electrophysiological recordings were performed in conjunction with behavioral phenotyping. Compared to mice models with classical depression or to schizophrenia models, the animal models with schizophrenia and depression comorbidity presented worse psychotic and depressive symptoms. These behavioral deficits are associated with impaired neuronal calcium activities in the frontal cortex and thalamic nuclei. Moreover, in sharp contrast to classical models that have a satisfactory response to antipsychotic or antidepressant drugs, this novel schizophrenia with depression model is resilient to combined drug treatment in terms of behavioral and functional recovery. Taken together, these data indicate that schizophrenia with depression likely involves a unique pathophysiology that is different from schizophrenia or depression alone.
Sigma-Aldrich
Picrotoxin, powder