Merck
  • Efficient and direct nucleophilic difluoromethylation of carbonyl compounds and imines with Me3SiCF2H at ambient or low temperature.
Efficient and direct nucleophilic difluoromethylation of carbonyl compounds and imines with Me3SiCF2H at ambient or low temperature.
Organic letters (2011-09-14)
Yanchuan Zhao, Weizhou Huang, Ji Zheng, Jinbo Hu
ABSTRACT
Since 1995, Me(3)SiCF(2)H has been widely believed to be an inefficient difluoromethylating agent, which requires harsh reaction conditions to cleave its rather inert Si-CF(2)H bond. However, it has now been found that, by using a proper Lewis base activator, Me(3)SiCF(2)H can efficiently difluoromethylate various aldehydes, ketones, and imines to give the corresponding products in good to excellent yields at room temperature or even at -78 °C.
Sigma-Aldrich
(Difluoromethyl)trimethylsilane, ≥98.0% (GC)