Merck
  • Rapid and sensitive determination of neomycin and kanamycin in measles, mumps, and rubella vaccine via high-performance liquid chromatography-tandem mass spectrometry using modified super-paramagnetic Fe3O4 nanospheres.
Rapid and sensitive determination of neomycin and kanamycin in measles, mumps, and rubella vaccine via high-performance liquid chromatography-tandem mass spectrometry using modified super-paramagnetic Fe3O4 nanospheres.
Journal of chromatography. A (2020-07-28)
Hamideh Hamidi, Mahshid Zarrineh, Ali Es-Haghi, Alireza Ghasempour
ABSTRACT
A simple magnetic dispersive solid-phase extraction (MDSPE) methodology based on mesoporous Fe3O4@ succinic acid nanospheres and high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) has been developed to determine kanamycin (KNM) and neomycin (NEO) contents in Measles, Mumps, and Rubella (MMR) vaccine products. The monodispersed mesoporous Fe3O4 nanospheres with self-assembled carboxyl terminated shell have been prepared via a simple solvothermal method. These as-synthesized mesoporous Fe3O4 nanospheres showed a high magnetic saturation value (Ms = 46 emu g-1) and large specific surface area (111.12 m2 g-1) which made them potential candidates as sorbents in magnetic solid-phase extraction. The adsorption experimental data fitted well with the Freundlich-Langmuir isotherm and followed a pseudo-second-order kinetic model. Moreover influential parameters on extraction efficiency were investigated and optimized. Under optimal conditions, the limits of detection for KNM and NEO were 1.0 and 0.1 ng mL-1, respectively. Recovery assessments using real samples exhibited recoveries in the range of 96.0 ± 4.3 to 101.5 ± 7.1 %, with relative standard deviations of <10.7% (for intra- day) and <14.6% (for inter- day). The proposed method was successfully applied for different spiked and un-spiked MMR vaccine samples. The presented extraction method provides a fast, selective, robust and practical platform for the detection of KNM and NEO in MMR vaccine samples.
Sigma-Aldrich
Heptafluorobutyric acid, 98%