Merck
  • Characterization and inactivation of the membrane-bound polyol dehydrogenase in Gluconobacter oxydans DSM 7145 reveals a role in meso-erythritol oxidation.
Characterization and inactivation of the membrane-bound polyol dehydrogenase in Gluconobacter oxydans DSM 7145 reveals a role in meso-erythritol oxidation.
Microbiology (Reading, England) (2010-03-13)
Jörn Voss, Armin Ehrenreich, Wolfgang Liebl
ABSTRACT
The growth of Gluconobacter oxydans DSM 7145 on meso-erythritol is characterized by two stages: in the first stage, meso-erythritol is oxidized almost stoichiometrically to L-erythrulose according to the Bertrand-Hudson rule. The second phase is distinguished from the first phase by a global metabolic change from membrane-bound meso-erythritol oxidation to L-erythrulose assimilation with concomitant accumulation of acetic acid. The membrane-associated erythritol-oxidizing enzyme was found to be encoded by a gene homologous to sldA known from other species of acetic acid bacteria. Disruption of this gene in the genome of G. oxydans DSM 7145 revealed that the membrane-bound polyol dehydrogenase not only oxidizes meso-erythritol but also has a broader substrate spectrum which includes C3-C6 polyols and D-gluconate and supports growth on these substrates. Cultivation of G. oxydans DSM 7145 on different substrates indicated that expression of the polyol dehydrogenase was not regulated, implying that the production of biomass of G. oxydans to be used as whole-cell biocatalysts in the biotechnological conversion of meso-erythritol to L-erythrulose, which is used as a tanning agent in the cosmetics industry, can be conveniently carried out with glucose as the growth substrate.
Sigma-Aldrich
meso-Erythritol, ≥99% (GC)