Merck
  • Assessment of the endocrine-disrupting effects of organophosphorus pesticide triazophos and its metabolites on endocrine hormones biosynthesis, transport and receptor binding in silico.
Assessment of the endocrine-disrupting effects of organophosphorus pesticide triazophos and its metabolites on endocrine hormones biosynthesis, transport and receptor binding in silico.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2019-08-20)
Fang-Wei Yang, Yi-Xuan Li, Fa-Zheng Ren, Jie Luo, Guo-Fang Pang
ABSTRACT
Triazophos (TAP) was a widely used organophosphorus insecticide in developing countries. TAP could produce specific metabolites triazophos-oxon (TAPO) and 1-phenyl-3-hydroxy-1,2,4-triazole (PHT) and non-specific metabolites diethylthiophosphate (DETP) and diethylphosphate (DEP). The objective of this study involved computational approaches to discover potential mechanisms of molecular interaction of TAP and its major metabolites with endocrine hormone-related proteins using molecular docking in silico. We found that TAP, TAPO and DEP showed high binding affinity with more proteins and enzymes than PHT and DETP. TAP might interfere with the endocrine function of the adrenal gland, and TAP might also bind strongly with glucocorticoid receptors and thyroid hormone receptors. TAPO might disrupt the normal binding of androgen receptor, estrogen receptor, progesterone receptor and adrenergic receptor to their natural hormone ligands. DEP might affect biosynthesis of steroid hormones and thyroid hormones. Meanwhile, DEP might disrupt the binding and transport of thyroid hormones in the blood and the normal binding of thyroid hormones to their receptors. These results suggested that TAP and DEP might have endocrine disrupting activities and were potential endocrine disrupting chemicals. Our results provided further reference for the comprehensive evaluation of toxicity of organophosphorus chemicals and their metabolites.
Sigma-Aldrich
Pyrrolidone hydrotribromide, 97%