Merck
  • First multi-reactive polysaccharide-based transurf to produce potentially biocompatible dextran-covered nanocapsules.
First multi-reactive polysaccharide-based transurf to produce potentially biocompatible dextran-covered nanocapsules.
Carbohydrate polymers (2019-09-02)
Laura Marcela Forero Ramirez, Jérôme Babin, Ariane Boudier, Caroline Gaucher, Marc Schmutz, Mériem Er-Rafik, Alain Durand, Jean-Luc Six, Cécile Nouvel
ABSTRACT
A multi-reactive polysaccharide-based transurf (acting both as macro-Chain Transfer Agent and stabilizer) was used to confine RAFT polymerization of methyl methacrylate (MMA) at the oil/water (o/w) miniemulsion interface. Dithiobenzoate groups and hydrophobic aliphatic side chains were introduced onto dextran, conferring it both transfer agent properties and ability to stabilize direct miniemulsion of MMA in the presence of a biocompatible oil, used as co-stabilizer. Because of their amphiphilic character, transurfs were initially adsorbed at the (o/w) interface and their reactive sites mediated RAFT polymerization via the R-group approach. PMMA-grafted dextran glycopolymers were consequently produced at the o/w interface, thus leading to dextran coverage/PMMA shell/oily core nanocapsules (NCs) as evidenced by Cryo-TEM analyses. The influence of dextran-based transurf chemistry and oil amount on MMA RAFT polymerization control was investigated. Positive preliminary results on NCs cytotoxicity suggest the potential of these objects for biomedical applications.
Sigma-Aldrich
2-Cyano-2-propyl benzodithioate, >97% (HPLC)