- Mast Cell-Derived Histamine Regulates Liver Ketogenesis via Oleoylethanolamide Signaling.
Mast Cell-Derived Histamine Regulates Liver Ketogenesis via Oleoylethanolamide Signaling.
The conversion of lipolysis-derived fatty acids into ketone bodies (ketogenesis) is a crucial metabolic adaptation to prolonged periods of food scarcity. The process occurs primarily in liver mitochondria and is initiated by fatty-acid-mediated stimulation of the ligand-operated transcription factor, peroxisome proliferator-activated receptor-α (PPAR-α). Here, we present evidence that mast cells contribute to the control of fasting-induced ketogenesis via a paracrine mechanism that involves secretion of histamine into the hepatic portal circulation, stimulation of liver H1 receptors, and local biosynthesis of the high-affinity PPAR-α agonist, oleoylethanolamide (OEA). Genetic or pharmacological interventions that disable any one of these events, including mast cell elimination, deletion of histamine- or OEA-synthesizing enzymes, and H1 blockade, blunt ketogenesis without affecting lipolysis. The results reveal an unexpected role for mast cells in the regulation of systemic fatty-acid homeostasis, and suggest that OEA may act in concert with lipolysis-derived fatty acids to activate liver PPAR-α and promote ketogenesis.