Merck
  • Planting carbon nanotubes onto supramolecular polymer matrices for waterproof non-contact self-healing.
Planting carbon nanotubes onto supramolecular polymer matrices for waterproof non-contact self-healing.
Nanoscale (2018-12-20)
Bo Li, Lei Kan, Shuai Zhang, Zhengdao Liu, Chao Li, Wei Li, Xinyue Zhang, Hao Wei, Ning Ma
ABSTRACT
Supramolecular polymers show unique and excellent properties due to the reversible and designable nature of the non-covalent interactions. Herein, ureido-pyrimidinone (UPy)-based supramolecular polymers were employed to fabricate the thermo-responsive composite materials with multi-walled carbon nanotubes (MWCNTs) by planting the MWCNTs onto the supramolecular polymer matrices via a simple surface spraying procedure. The MWCNTs coating on the surface of the supramolecular polymer matrices gave the composite film superhydrophobic and conductive properties, and it had a non-contact healable ability underwater under 808 nm near-infrared light (NIR) irradiation. Moreover, the UPy-based supramolecular polymers acted as thermo-responsive matrices to guarantee the self-healing properties at a relatively low temperature, such as body temperature (33 °C-34 °C). The supramolecular polymer/MWCNTs composite materials exhibited excellent strain sensitivities and could be used to prepare human motion-monitoring devices. This line of research may find a promising practical application in healable wearable devices used in underwater environments.