Merck
  • Amygdala NPY Circuits Promote the Development of Accelerated Obesity under Chronic Stress Conditions.
Amygdala NPY Circuits Promote the Development of Accelerated Obesity under Chronic Stress Conditions.
Cell metabolism (2019-04-30)
Chi Kin Ip, Lei Zhang, Aitak Farzi, Yue Qi, Ireni Clarke, Felicia Reed, Yan-Chuan Shi, Ronaldo Enriquez, Chris Dayas, Bret Graham, Denovan Begg, Jens C Brüning, Nicola J Lee, Diana Hernandez-Sanchez, Gopana Gopalasingam, Julia Koller, Ramon Tasan, Günther Sperk, Herbert Herzog
ABSTRACT
Neuropeptide Y (NPY) exerts a powerful orexigenic effect in the hypothalamus. However, extra-hypothalamic nuclei also produce NPY, but its influence on energy homeostasis is unclear. Here we uncover a previously unknown feeding stimulatory pathway that is activated under conditions of stress in combination with calorie-dense food; NPY neurons in the central amygdala are responsible for an exacerbated response to a combined stress and high-fat-diet intervention. Central amygdala NPY neuron-specific Npy overexpression mimics the obese phenotype seen in a combined stress and high-fat-diet model, which is prevented by the selective ablation of Npy. Using food intake and energy expenditure as readouts, we demonstrate that selective activation of central amygdala NPY neurons results in increased food intake and decreased energy expenditure. Mechanistically, it is the diminished insulin signaling capacity on central amygdala NPY neurons under combined stress and high-fat-diet conditions that leads to the exaggerated development of obesity.
Sigma-Aldrich
Monoclonal Anti-NPY antibody produced in mouse, clone 3B5, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Clozapine N-oxide
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)