Merck
  • A senescence program controlled by p53 and p16INK4a contributes to the outcome of cancer therapy.
A senescence program controlled by p53 and p16INK4a contributes to the outcome of cancer therapy.
Cell (2002-05-23)
Clemens A Schmitt, Jordan S Fridman, Meng Yang, Soyoung Lee, Eugene Baranov, Robert M Hoffman, Scott W Lowe
ABSTRACT
p53 and INK4a/ARF mutations promote tumorigenesis and drug resistance, in part, by disabling apoptosis. We show that primary murine lymphomas also respond to chemotherapy by engaging a senescence program controlled by p53 and p16(INK4a). Hence, tumors with p53 or INK4a/ARF mutations-but not those lacking ARF alone-respond poorly to cyclophosphamide therapy in vivo. Moreover, tumors harboring a Bcl2-mediated apoptotic block undergo a drug-induced cytostasis involving the accumulation of p53, p16(INK4a), and senescence markers, and typically acquire p53 or INK4a mutations upon progression to a terminal stage. Finally, mice bearing tumors capable of drug-induced senescence have a much better prognosis following chemotherapy than those harboring tumors with senescence defects. Therefore, cellular senescence contributes to treatment outcome in vivo.
Sigma-Aldrich
Cellular Senescence Assay, Cellular Senescence Assay Kits provide all the reagents required to efficiently detect SA-β-gal activity at pH 6.0 in cultured cells & tissue sections.