Merck
  • On the use of L-012, a luminol-based chemiluminescent probe, for detecting superoxide and identifying inhibitors of NADPH oxidase: a reevaluation.
On the use of L-012, a luminol-based chemiluminescent probe, for detecting superoxide and identifying inhibitors of NADPH oxidase: a reevaluation.
Free radical biology & medicine (2013-10-02)
Jacek Zielonka, J David Lambeth, Balaraman Kalyanaraman
ABSTRACT
L-012, a luminol-based chemiluminescent (CL) probe, is widely used in vitro and in vivo to detect NADPH oxidase (Nox)-derived superoxide (O2(*-)) and identify Nox inhibitors. Yet understanding of the free radical chemistry of the L-012 probe is still lacking. We report that peroxidase and H2O2 induce superoxide dismutase (SOD)-sensitive, L-012-derived CL in the presence of oxygen. O2(*-) alone does not react with L-012 to emit luminescence. Self-generated O2(*-) during oxidation of L-012 and luminol analogs artifactually induce CL inhibitable by SOD. These aspects make assays based on luminol analogs less than ideal for specific detection and identification of O2(*-) and NOX inhibitors.
Sigma-Aldrich
L-012 sodium salt, ≥98% (HPLC)