Merck
  • A virtual screening hit reveals new possibilities for developing group III metabotropic glutamate receptor agonists.
A virtual screening hit reveals new possibilities for developing group III metabotropic glutamate receptor agonists.
Journal of medicinal chemistry (2010-03-12)
Chelliah Selvam, Nadia Oueslati, Isabelle A Lemasson, Isabelle Brabet, Delphine Rigault, Tiphanie Courtiol, Sara Cesarini, Nicolas Triballeau, Hugues-Olivier Bertrand, Cyril Goudet, Jean-Philippe Pin, Francine C Acher
ABSTRACT
(R)-PCEP (3-amino-3-carboxypropyl-2'-carboxyethyl phosphinic acid, 1), a new metabotropic glutamate receptor 4 (mGlu4R) agonist, was discovered in a previously reported virtual screening. The (S)-enantiomer and a series of derivatives were synthesized and tested on recombinant mGlu4 receptors. A large number of derivatives activated this receptor but was not able to discriminate between mGlu4 and mGlu8 receptors. The most potent ones 6 and 12 displayed an EC(50) of 1.0 +/- 0.2 microM at mGlu4R. Interestingly these agonists with longer alkyl chains revealed a new binding pocket adjacent to the glutamate binding site, which is lined with residues that differ among the mGluR subtypes and that will allow the design of more selective compounds. Additionally 6 was able to activate mGlu7 receptor with an EC(50) of 43 +/- 16 microM and is thus significantly more potent than L-AP4 (EC(50) of 249 +/- 106 microM).
Sigma-Aldrich
L-(+)-2-Amino-4-phosphonobutyric acid, optical purity optical purity: ≥95% (HPLC, Marfey′s reagent)