Merck
  • Facilely Synthesized spiro[fluorene-9,9'-phenanthren-10'-one] in Donor-Acceptor-Donor Hole-Transporting Materials for Perovskite Solar Cells.
Facilely Synthesized spiro[fluorene-9,9'-phenanthren-10'-one] in Donor-Acceptor-Donor Hole-Transporting Materials for Perovskite Solar Cells.
ChemSusChem (2018-07-08)
Yih-Chun Chen, Shao-Ku Huang, Shao-Sian Li, Yao-Yu Tsai, Chih-Ping Chen, Chun-Wei Chen, Yuan Jay Chang
ABSTRACT
We have demonstrated two novel donor-acceptor-donor (D-A-D) hole-transport material (HTM) with spiro[fluorene-9,9'-phenanthren-10'-one] as the core structure, which can be synthesized through a low-cost process in high yield. Compared to the incorporation of the conventional HTM of commonly used 2,2',7,7'-tetrakis[N,N-di(4-methoxyphenyl)amino]-9,9'-spirobifluorene (Spiro-OMeTAD), the synthesis process is greatly simplified for the presented D-A-D materials, including a minimum number of purification processes. This results in an increased production yield (>55 %) and suppressed production cost (<30 $ g-1 ), in addition to high power conversion efficiency (PCE) in perovskite solar cells (PSCs). The PCE of a PSC using our D-A-D HTM reaches 16.06 %, similar to that of Spiro-OMeTAD (16.08 %), which is attributed to comparable hole mobility and charge-transfer efficiency. D-A-D HTMs also provide better moisture resistivity to prolong the lifetime of PSCs under ambient conditions relative to their Spiro-OMeTAD counterparts. The proposed new type of D-A-D HTM has shown promising performance as an alternative HTM for PSCs and can be synthesized with high production throughput.
Sigma-Aldrich
Tri(o-tolyl)phosphine, 97%