Merck
  • Broadband Sensitization of Lanthanide Emission with Indium Phosphide Quantum Dots for Visible to Near-Infrared Downshifting.
Broadband Sensitization of Lanthanide Emission with Indium Phosphide Quantum Dots for Visible to Near-Infrared Downshifting.
Journal of the American Chemical Society (2018-06-21)
Joseph K Swabeck, Stefan Fischer, Noah D Bronstein, A Paul Alivisatos
ABSTRACT
Semiconductor quantum dot (QD)-sensitized lanthanide ions hold great promise in producing a broadly absorbing and sharply emitting luminophore, but their synthesis has proven to be difficult. We report the first synthesis of core/shell/shell InP/Ln xY1- xF3/ShF3 (Ln = Yb, Nd; Sh = Lu, Y) nanocrystals that exhibit a broad visible absorption coupled to a sharp near-infrared emission. Additionally, this is the first report of Nd being coupled to a QD absorber. We characterize the system with a variety of electron microscopy and X-ray techniques that prove this unique structure. Optical measurements confirm the correlation of the Ln3+ emission to the QD absorption, while the presence of a trap-state emission gives a clue as to the mechanism of energy transfer between the dot and the lanthanide.
Sigma-Aldrich
Ytterbium(III) oxide, 99.9% trace metals basis
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%