- Endocrine disruption in Chinese rare minnow (Gobiocypris rarus) after long-term exposure to low environmental concentrations of progestin megestrol acetate.
Endocrine disruption in Chinese rare minnow (Gobiocypris rarus) after long-term exposure to low environmental concentrations of progestin megestrol acetate.
Synthetic progestins are widely used pharmaceutical agents that have become common contaminants in the aquatic environment. The potential adverse effects of long-term exposure on aquatic wildlife, however, are not fully understood. The aim of this study was to investigate the endocrine disruption in Chinese rare minnow (Gobiocypris rarus) in response to megestrol acetate (MTA) exposure. Newly-hatched Chinese rare minnow larvae were exposed to MTA at a nominal concentration of either 1 ng/L (detected concentrations ranged from 0.18 to 0.93 ng/L) or 10 ng/L (detected concentrations ranged from 4.27 to 9.64 ng/L) for 6 months and the effects on growth, sex steroid hormones, gonadal histology, and steroidogenic genes expression were determined. After 6 months of exposure to a nominal concentration of 10 ng/L MTA, the body weight and condition factors were significantly increased in fish of both sexes. Exposure to a nominal concentration of 10 ng/L MTA significantly reduced plasma concentrations of estradiol and 11-ketotestosterone in female fish while also reducing testosterone and 11-ketotestosterone in male fish. Gonad histology revealed significantly reduced proportions of vitellogenic oocytes in female fish exposed to a nominal concentration of 10 ng/L MTA and induction of atretic follicles in female fish exposed to both nominal concentrations of MTA. The expression of cyp19a1a and cyp17a1 in the gonads was up-regulated in the ovaries while down-regulated in the testes. Our results indicate that MTA can induce endocrine disruption in Chinese rare minnow at the low concentrations found in contaminated environments. This indicates a potentially high ecological risk from MTA to fish populations in MTA-contaminated aquatic environments in China and may also in other regions.