Merck
  • Succinobucol, a Lipid-Lowering Drug, Protects Against 3-Nitropropionic Acid-Induced Mitochondrial Dysfunction and Oxidative Stress in SH-SY5Y Cells via Upregulation of Glutathione Levels and Glutamate Cysteine Ligase Activity.
Succinobucol, a Lipid-Lowering Drug, Protects Against 3-Nitropropionic Acid-Induced Mitochondrial Dysfunction and Oxidative Stress in SH-SY5Y Cells via Upregulation of Glutathione Levels and Glutamate Cysteine Ligase Activity.
Molecular neurobiology (2015-01-27)
Dirleise Colle, Danúbia Bonfanti Santos, Juliana Montagna Hartwig, Marcelo Godoi, Daiane Fátima Engel, Andreza Fabro de Bem, Antonio L Braga, Marcelo Farina
ABSTRACT
Succinobucol (succinyl ester of probucol) is a lipid-lowering compound with anti-inflammatory and antioxidant properties. Recent experimental evidence has highlighted the potential neuroprotective effects of succinobucol. In the present study, cultured neuroblastoma (SH-SY5Y) cells were used to investigate mechanisms mediating the potential protective effect of succinobucol against mitochondrial metabolic impairment and oxidative stress induced by 3-nitropropionic acid (3-NP), a succinate dehydrogenase inhibitor that has been used in experimental models of the Huntington disease (HD). 3-NP decreased cellular viability after 24 h of incubation. This decline in cellular viability was preceded by (i) reduced mitochondrial complex II activity, (ii) increased reactive species generation, (iii) decreased mitochondrial membrane potential (ΔΨm), and (iv) diminished glutathione (GSH) levels. Succinobucol pretreatment (6 days) significantly prevented 3-NP-induced loss of cellular viability, generation of reactive oxygen species, and decrease of ΔΨm. However, succinobucol pretreatment did not protect against 3-NP-induced inhibition of mitochondrial complex II activity, pointing to the mitigation of secondary events resultant from mitochondrial complex II inhibition. Succinobucol pretreatment (6 days) significantly increased (50 %) the levels of GSH in SH-SY5Y cells, and this event was paralleled by significant increases in glutamate cysteine ligase messenger RNA (mRNA) expression and activity (GCL; the first enzyme in the GSH biosynthesis). The present findings are the first to show that succinobucol increases GSH levels via upregulation of GCL activity (possibly through the activation of the nuclear (erythroid-derived 2)-related factor (Nrf2)/antioxidant response element (ARE) pathway), displaying protective effects against mitochondrial dysfunction-derived oxidative stress.
Sigma-Aldrich
Succinobucol, ≥98% (HPLC)