Merck
  • Effects of NK1 receptors on gastric motor functions and satiation in healthy humans: results from a controlled trial with the NK1 antagonist aprepitant.
Effects of NK1 receptors on gastric motor functions and satiation in healthy humans: results from a controlled trial with the NK1 antagonist aprepitant.
American journal of physiology. Gastrointestinal and liver physiology (2017-08-18)
Deepti Jacob, Irene Busciglio, Duane Burton, Houssam Halawi, Ibironke Oduyebo, Deborah Rhoten, Michael Ryks, W Scott Harmsen, Michael Camilleri
ABSTRACT
Aprepitant, an NK1 receptor antagonist, is approved for the treatment of chemotherapy-induced or postoperative emesis by blocking NK1 receptors in the brain stem vomiting center. The effects of NK1 receptors on gastric functions and postprandial symptoms in humans are unclear; a single, crossover study did not show a significant effect of aprepitant on gastrointestinal transit. Our aim was to compare, in a randomized, double-blind, placebo-controlled, parallel-group study (12 healthy volunteers per group), the effects of aprepitant vs. placebo on gastric emptying of solids (by scintigraphy) with a 320-kcal meal, gastric volumes (GVs; fasting and accommodation by single photon emission-computed tomography ), satiation [maximum tolerated volume (MTV)], and symptoms after a dyspeptogenic meal of Ensure. Aprepitant (125 mg on
Sigma-Aldrich
Aprepitant, ≥98% (HPLC)