Skip to Content
Merck
TSKgel® SuperAW5000 HPLC Column phase hydroxylated methacrylate, L × I.D. 15 cm × 6 mm, 7 μm particle size

TSKgel® SuperAW5000 (7 µm) HPLC Columns

TSKgel® SuperAW5000 HPLC Column phase hydroxylated methacrylate, L × I.D. 15 cm × 6 mm, 7 μm particle size
Synonym(s):
TSKgel® Size Exclusion (PW-Type) HPLC Column

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1