Skip to Content
Merck
Sunset Yellow FCF analytical standard

Sunset Yellow FCF

Sunset Yellow FCF analytical standard
Synonym(s):
Food Yellow 3, Orange Yellow S
Empirical Formula (Hill Notation):
C16H10N2Na2O7S2
CAS Number:
Molecular Weight:
452.37
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing