Skip to Content
Merck
S-Nitrosoglutathione ≥97%

S-Nitrosoglutathione

S-Nitrosoglutathione ≥97%
Synonym(s):
GSNO, SNOG, S-Nitrosoglutathione
Empirical Formula (Hill Notation):
C10H16N4O7S
CAS Number:
Molecular Weight:
336.32

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2