
Ochratoxin A-d5 solution

Synonym(s):
Ochratoxin A-(phenyl-d5) solution
Empirical Formula (Hill Notation):
C20D5H13ClNO6
CAS Number:
Molecular Weight:
408.84
Products
Product Category
Brand
Formula Weight
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing