Skip to Content
Merck
Nonacosane analytical standard

Nonacosane

Nonacosane analytical standard
Linear Formula:
CH3(CH2)27CH3
CAS Number:
Molecular Weight:
408.79
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing