
Monoclonal Anti-E2F6 antibody produced in mouse

Synonym(s):
Anti-E2F transcription factor 6, Anti-E2F-6
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing